Search
Search
| Gene Symbol: PML | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206503.1 | NP_001193432.1 | 3 - 4 | 1275 | 98 | |
| GenBank mRNA | BC149786.1 | - | 3 - 4 | 1236 | 98 | |
| HQ402594.1 | - | 2 - 3 | 979 | 98 | ||
Set Membership: |
|