Search
Search

COG3| Gene Symbol: COG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206537.1 | NP_001193466.1 | 19 - 20 | 2277 | 61 | |
| GenBank mRNA | EE903906.1 | - | 2 - 3 | 198 | 61 | |
| EE959930.1 | - | 1 - 2 | 121 | 61 | ||
Set Membership: |
|