Search
Search

DCAF12| Gene Symbol: DCAF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167896.2 | NP_001161368.1 | 6 - 7 | 1171 | 83 | |
| GenBank mRNA | BC105241.1 | - | 6 - 7 | 862 | 83 | |
| BC150062.1 | - | 6 - 7 | 1170 | 83 | ||
Set Membership: |
|