Search
Search

KLF10| Gene Symbol: KLF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168462.1 | NP_001161934.1 | 3 - 4 | 1309 | 71 | |
| GenBank mRNA | AB530320.1 | - | 3 - 4 | 1309 | 71 | |
| BC149823.1 | - | 3 - 4 | 1234 | 71 | ||
Set Membership: |
|