Search
Search

HSF4| Gene Symbol: HSF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191202.1 | NP_001178131.1 | 12 - 13 | 1399 | 67 | |
| GenBank mRNA | BC149044.1 | - | 11 - 12 | 1543 | 67 | |
| CK773361.1 | - | 1 - 2 | 311 | 67 | ||
Set Membership: |
|