Search
Search
| Gene Symbol: RGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173957.2 | NP_776382.1 | 6 - 7 | 947 | 99 | |
| GenBank mRNA | AB035446.1 | - | 6 - 7 | 947 | 99 | |
| BC103461.1 | - | 6 - 7 | 959 | 99 | ||
Set Membership: |
|