Search
Search

BOD1L1| Gene Symbol: BOD1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192190.3 | NP_001179119.2 | 25 - 26 | 8974 | 70 | |
| GenBank mRNA | BC149869.1 | - | 16 - 17 | 1247 | 70 | |
Set Membership: |
|