Search
Search

NDOR1| Gene Symbol: NDOR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046158.2 | NP_001039623.2 | 7 - 8 | 931 | 56 | |
| GenBank mRNA | BC112851.1 | - | 7 - 8 | 931 | 56 | |
| BT025363.1 | - | 7 - 8 | 859 | 56 | ||
Set Membership: |
|