Search
Search

TXNDC16| Gene Symbol: TXNDC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205394.1 | NP_001192323.1 | 14 - 15 | 1776 | 71 | |
| GenBank mRNA | BC123407.1 | - | 14 - 15 | 1776 | 71 | |
Set Membership: |
|