Search
Search

rbc-1| Gene Symbol: rbc-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270317.4 | NP_001257246.2 | 17 - 18 | 4329 | 96 | |
| NM_001373583.1 | NP_001360775.1 | 17 - 18 | 4296 | 96 | ||
| NM_001381151.1 | NP_001368758.1 | 17 - 18 | 4312 | 96 | ||
| NM_001381152.1 | NP_001368759.1 | 17 - 18 | 4312 | 96 | ||
| NM_001381153.1 | NP_001368756.1 | 17 - 18 | 4279 | 96 | ||
| NM_001381154.1 | NP_001368757.1 | 17 - 18 | 4279 | 96 | ||
| NM_001381155.1 | NP_001368146.1 | 17 - 18 | 4306 | 96 | ||
| NM_001381156.2 | NP_001368760.1 | 17 - 18 | 4302 | 96 | ||
Set Membership: |
|