Search
Search

tbh-1| Gene Symbol: tbh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270335.2 | NP_001257264.1 | 4 - 5 | 607 | 69 | |
| NM_001270336.3 | NP_001257265.1 | 3 - 4 | 404 | 69 | ||
| NM_001313436.5 | NP_001300365.1 | 5 - 6 | 554 | 69 | ||
Set Membership: |
|