Search
Search

eat-20| Gene Symbol: eat-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029570.6 | NP_001024741.1 | 11 - 12 | 1479 | 104 | |
| NM_001383685.1 | 12 - 13 | 1531 | 104 | |||
| NR_002425.1 | 11 - 12 | 1381 | 104 | |||
| GenBank mRNA | AB032748.1 | - | 11 - 12 | 1477 | 104 | |
| AB032749.1 | - | 11 - 12 | 1477 | 104 | ||
Set Membership: |
|