Search
Search

eat-20| Gene Symbol: eat-20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029570.6 | NP_001024741.1 | 9 - 10 | 1253 | 71 | |
| NM_001383685.1 | 10 - 11 | 1305 | 71 | |||
| GenBank mRNA | AB032748.1 | - | 9 - 10 | 1251 | 71 | |
| AB032749.1 | - | 9 - 10 | 1251 | 71 | ||
Set Membership: |
|