Search
Search

aak-2| Gene Symbol: aak-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029697.5 | NP_001024868.1 | 6 - 7 | 1169 | 66 | |
| NM_078309.7 | NP_510710.2 | 7 - 8 | 1318 | 66 | ||
| NM_078310.7 | NP_510711.2 | 7 - 8 | 1314 | 66 | ||
| GenBank mRNA | AY347273.1 | - | 7 - 8 | 1312 | 66 | |
Set Membership: |
|