Search
Search

nhr-23| Gene Symbol: nhr-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025804.5 | NP_001020975.1 | 3 - 4 | 729 | 68 | |
| NM_001025806.6 | NP_001020977.1 | 2 - 3 | 349 | 68 | ||
| NM_001037880.5 | NP_001032969.1 | 1 - 2 | 98 | 68 | ||
| NM_001037882.5 | NP_001032971.1 | 2 - 3 | 485 | 68 | ||
| NM_001380645.1 | NP_001367012.1 | 2 - 3 | 498 | 68 | ||
| NM_001380646.1 | NP_001367128.1 | 2 - 3 | 487 | 68 | ||
| GenBank mRNA | AF332203.1 | - | 3 - 4 | 640 | 68 | |
Set Membership: |
|