Search
Search

scpl-1| Gene Symbol: scpl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128939.2 | NP_001122411.1 | 6 - 7 | 691 | 111 | |
| NM_001383711.1 | 10 - 11 | 1052 | 111 | |||
| NM_170918.6 | NP_740912.2 | 9 - 10 | 1463 | 111 | ||
Set Membership: |
|