Search
Search

pmr-1| Gene Symbol: pmr-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026691.5 | NP_001021862.1 | 9 - 10 | 2583 | 65 | |
| NM_001373648.3 | NP_001359606.1 | 9 - 10 | 2609 | 65 | ||
| GenBank mRNA | AJ303081.1 | - | 9 - 10 | 2637 | 65 | |
| AJ303082.1 | - | 9 - 10 | 2631 | 65 | ||
Set Membership: |
|