Search
Search

kin-1| Gene Symbol: kin-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_061204.4 | NP_493605.1 | 6 - 7 | 1053 | 65 | |
| NM_170958.5 | NP_740954.1 | 8 - 9 | 1182 | 65 | ||
| NM_170960.6 | NP_740956.1 | 6 - 7 | 957 | 65 | ||
| NM_170962.5 | NP_740958.1 | 6 - 7 | 1124 | 65 | ||
| NM_170964.6 | NP_740960.1 | 6 - 7 | 1129 | 65 | ||
| NM_170966.3 | NP_740962.1 | 6 - 7 | 1259 | 65 | ||
Set Membership: |
|