Search
Search

dnj-5| Gene Symbol: dnj-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381467.2 | NP_001367422.1 | 11 - 12 | 2385 | 134 | |
| NM_062999.9 | NP_495400.2 | 11 - 12 | 2372 | 134 | ||
| NM_171014.6 | NP_741015.1 | 11 - 12 | 2369 | 134 | ||
| NM_171015.6 | NP_741016.1 | 11 - 12 | 2356 | 134 | ||
Set Membership: |
|