Search
Search

npp-21| Gene Symbol: npp-21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001373741.2 | NP_001360471.1 | 4 - 5 | 1438 | 70 | |
| NM_001373742.2 | NP_001360472.1 | 4 - 5 | 1438 | 70 | ||
| NM_001373743.2 | NP_001360024.1 | 4 - 5 | 1438 | 70 | ||
| NM_001373744.2 | NP_001360023.1 | 4 - 5 | 1440 | 70 | ||
| NR_151568.1 | 4 - 5 | 1438 | 70 | |||
Set Membership: |
|