Search
Search

unc-53| Gene Symbol: unc-53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026998.5 | NP_001022169.1 | 18 - 19 | 2805 | 99 | |
| NM_001026999.8 | NP_001022170.1 | 20 - 21 | 3141 | 99 | ||
| NM_001027000.6 | NP_001022171.1 | 17 - 18 | 2936 | 99 | ||
| NM_001027001.5 | NP_001022172.1 | 12 - 13 | 1881 | 99 | ||
| NM_001267342.3 | NP_001254271.1 | 18 - 19 | 2805 | 99 | ||
| NM_001267343.3 | NP_001254272.1 | 19 - 20 | 3018 | 99 | ||
| NM_001267344.3 | NP_001254273.1 | 13 - 14 | 2217 | 99 | ||
| NM_171039.3 | NP_741045.1 | 19 - 20 | 2969 | 99 | ||
Set Membership: |
|