Search
Search

pef-1| Gene Symbol: pef-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267907.3 | NP_001254836.1 | 2 - 3 | 237 | 81 | |
| NM_171080.6 | NP_741091.1 | 3 - 4 | 432 | 81 | ||
| GenBank mRNA | AF023454.1 | - | 3 - 4 | 460 | 81 | |
Set Membership: |
|