Search
Search

frm-8| Gene Symbol: frm-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065271.6 | NP_497672.2 | 9 - 10 | 2073 | 94 | |
| NM_171096.6 | NP_741109.1 | 10 - 11 | 2197 | 94 | ||
| NM_171097.5 | NP_741110.1 | 9 - 10 | 2085 | 94 | ||
Set Membership: |
|