Search
Search

ten-1| Gene Symbol: ten-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027552.5 | NP_001022723.2 | 10 - 11 | 3467 | 137 | |
| NM_001268011.3 | NP_001254940.1 | 13 - 14 | 3861 | 137 | ||
| NM_001268012.3 | NP_001254941.1 | 11 - 12 | 3639 | 137 | ||
| NM_001379777.2 | NP_001367791.1 | 10 - 11 | 3473 | 137 | ||
| GenBank mRNA | AB206835.1 | - | 10 - 11 | 3467 | 137 | |
| AB206836.1 | - | 5 - 6 | 2973 | 137 | ||
Set Membership: |
|