Search
Search

soc-2| Gene Symbol: soc-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026088.4 | NP_001021259.1 | 3 - 4 | 553 | 70 | |
| NM_001306829.3 | NP_001293758.1 | 4 - 5 | 706 | 70 | ||
| NM_001306830.3 | NP_001293759.1 | 4 - 5 | 497 | 70 | ||
| NM_171332.5 | NP_741391.2 | 4 - 5 | 471 | 70 | ||
| GenBank mRNA | AF054827.1 | - | 3 - 4 | 560 | 70 | |
| AF068919.1 | - | 3 - 4 | 466 | 70 | ||
Set Membership: |
|