Search
Search

lin-45| Gene Symbol: lin-45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306912.2 | NP_001293841.1 | 4 - 5 | 779 | 80 | |
| NM_001306913.3 | NP_001293842.1 | 5 - 6 | 845 | 80 | ||
| NM_001306914.2 | NP_001293843.1 | 4 - 5 | 743 | 80 | ||
| NM_001380288.1 | NP_001368430.1 | 3 - 4 | 544 | 80 | ||
| NM_001383109.1 | 5 - 6 | 802 | 80 | |||
| NM_171367.8 | NP_741430.3 | 4 - 5 | 676 | 80 | ||
| GenBank mRNA | AY455928.1 | - | 4 - 5 | 738 | 80 | |
| AY493413.1 | - | 4 - 5 | 730 | 80 | ||
| AY493414.1 | - | 3 - 4 | 559 | 80 | ||
Set Membership: |
|