Search
Search

ctg-1| Gene Symbol: ctg-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276522.3 | NP_001263451.1 | 4 - 5 | 617 | 121 | |
| NM_001276523.3 | NP_001263452.1 | 3 - 4 | 470 | 121 | ||
| NM_059592.6 | NP_491993.1 | 6 - 7 | 1179 | 121 | ||
Set Membership: |
|