Search
Search

rrf-1| Gene Symbol: rrf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001263626.3 | NP_001250555.1 | 14 - 15 | 3436 | 116 | |
| NM_001263627.3 | NP_001250556.1 | 13 - 14 | 3183 | 116 | ||
| GenBank mRNA | AF159144.1 | - | 14 - 15 | 3436 | 116 | |
Set Membership: |
|