Search
Search

rbg-2| Gene Symbol: rbg-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026460.3 | NP_001021631.1 | 10 - 11 | 3157 | 71 | |
| NM_001026461.4 | NP_001021632.1 | 10 - 11 | 3155 | 71 | ||
Set Membership: |
|