Search
Search

tlf-1| Gene Symbol: tlf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001129002.4 | NP_001122474.1 | 3 - 4 | 285 | 63 | |
| NM_059955.7 | NP_492356.1 | 3 - 4 | 274 | 63 | ||
| GenBank mRNA | AF228692.1 | - | 3 - 4 | 296 | 63 | |
Set Membership: |
|