Search
Search

acr-19| Gene Symbol: acr-19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136284.4 | NP_001129756.1 | 3 - 4 | 432 | 74 | |
| NM_060028.7 | NP_492429.2 | 3 - 4 | 433 | 74 | ||
| GenBank mRNA | AY523514.1 | - | 3 - 4 | 433 | 74 | |
Set Membership: |
|