Search
Search

lin-41| Gene Symbol: lin-41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025827.5 | NP_001020998.1 | 2 - 3 | 329 | 64 | |
| NM_001025828.6 | NP_001020999.1 | 2 - 3 | 329 | 64 | ||
| GenBank mRNA | AF195610.1 | - | 1 - 2 | 134 | 64 | |
| AF195611.1 | - | 1 - 2 | 134 | 64 | ||
Set Membership: |
|