Search
Search

ced-12| Gene Symbol: ced-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306347.3 | NP_001293276.1 | 6 - 7 | 1850 | 106 | |
| NM_060292.7 | NP_492693.1 | 6 - 7 | 1813 | 106 | ||
| GenBank mRNA | AF324505.1 | - | 6 - 7 | 1834 | 106 | |
| AF416781.1 | - | 6 - 7 | 1815 | 106 | ||
Set Membership: |
|