Search
Search

ced-12| Gene Symbol: ced-12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306347.3 | NP_001293276.1 | 7 - 8 | 2091 | 83 | |
| NM_060292.7 | NP_492693.1 | 7 - 8 | 2054 | 83 | ||
| GenBank mRNA | AF324505.1 | - | 7 - 8 | 2075 | 83 | |
| AF416781.1 | - | 7 - 8 | 2056 | 83 | ||
Set Membership: |
|