Search
Search

sptf-1| Gene Symbol: sptf-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026294.3 | NP_001021465.1 | 2 - 3 | 330 | 77 | |
| NM_001373640.1 | NP_001359696.1 | 2 - 3 | 330 | 77 | ||
Set Membership: |
|