Search
Search

atm-1| Gene Symbol: atm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306285.2 | NP_001293214.1 | 10 - 11 | 2446 | 157 | |
| NM_001306286.3 | NP_001293215.1 | 1 - 2 | 403 | 157 | ||
Set Membership: |
|