Search
Search

grh-1| Gene Symbol: grh-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306482.4 | NP_001293411.1 | 3 - 4 | 947 | 113 | |
| NM_058436.5 | NP_490837.2 | 3 - 4 | 948 | 113 | ||
| GenBank mRNA | AY323527.1 | - | 3 - 4 | 947 | 113 | |
Set Membership: |
|