Search
Search

dyb-1| Gene Symbol: dyb-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001383732.1 | 1 - 2 | 144 | 91 | ||
| NM_058459.6 | NP_490860.1 | 1 - 2 | 151 | 91 | ||
| GenBank mRNA | AJ131742.1 | - | 1 - 2 | 128 | 91 | |
Set Membership: |
|