Search
Search

bre-4| Gene Symbol: bre-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_058471.2 | NP_490872.1 | 3 - 4 | 587 | 87 | |
| GenBank mRNA | AY130767.1 | - | 2 - 3 | 489 | 87 | |
| AY533306.1 | - | 2 - 3 | 489 | 87 | ||
Set Membership: |
|