Search
Search

afd-1| Gene Symbol: afd-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026489.7 | NP_001021660.2 | 10 - 11 | 1491 | 66 | |
| NM_001026490.6 | NP_001021661.2 | 10 - 11 | 1489 | 66 | ||
| NM_001306400.4 | NP_001293329.1 | 10 - 11 | 1493 | 66 | ||
| NM_001306401.3 | NP_001293330.1 | 5 - 6 | 628 | 66 | ||
| NM_001306402.3 | NP_001293331.1 | 5 - 6 | 628 | 66 | ||
| NM_001306403.3 | NP_001293332.1 | 4 - 5 | 442 | 66 | ||
| NM_001306404.3 | NP_001293333.1 | 4 - 5 | 442 | 66 | ||
| NM_001306405.3 | NP_001293334.1 | 1 - 2 | 163 | 66 | ||
| NM_001306406.3 | NP_001293335.1 | 1 - 2 | 163 | 66 | ||
| NM_001356800.4 | NP_001343732.1 | 7 | 1044 | 66 | ||
| NM_058537.5 | NP_490938.3 | 10 - 11 | 1485 | 66 | ||
Set Membership: |
|