Search
Search

lgc-51| Gene Symbol: lgc-51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306430.1 | NP_001293359.1 | 3 - 4 | 446 | 64 | |
| NM_001306431.1 | NP_001293360.1 | 3 - 4 | 446 | 64 | ||
Set Membership: |
|