Search
Search

nekl-1| Gene Symbol: nekl-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306391.3 | NP_001293320.1 | 11 - 12 | 1591 | 112 | |
| NM_001306392.3 | NP_001293321.1 | 10 - 11 | 1477 | 112 | ||
| NR_131355.1 | 11 - 12 | 1591 | 112 | |||
| NR_131356.1 | 10 - 11 | 1480 | 112 | |||
Set Membership: |
|