Search Thermo Fisher Scientific
Gene Symbol: larp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058808.5 | NP_491209.3 | 6 - 7 | 1003 | 75 | |
NM_182024.8 | NP_871824.3 | 6 - 7 | 997 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |