Search
Search

unc-73| Gene Symbol: unc-73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026325.2 | NP_001021496.1 | 18 - 19 | 4910 | 66 | |
| NM_001026326.4 | NP_001021497.1 | 18 - 19 | 4798 | 66 | ||
| NM_001026327.4 | NP_001021498.1 | 2 - 3 | 444 | 66 | ||
| NM_001262584.2 | NP_001249513.1 | 2 - 3 | 491 | 66 | ||
| NM_001375256.3 | NP_001361940.1 | 2 - 3 | 238 | 66 | ||
| NM_001381834.2 | NP_001367474.1 | 18 - 19 | 4798 | 66 | ||
| GenBank mRNA | AF048834.1 | - | 18 - 19 | 4910 | 66 | |
| AF048835.1 | - | 18 - 19 | 4910 | 66 | ||
Set Membership: |
|