Search
Search

unc-89| Gene Symbol: unc-89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025813.2 | NP_001020984.1 | 31 - 32 | 19702 | 71 | |
| NM_001025814.4 | NP_001020985.1 | 31 - 32 | 19511 | 71 | ||
| NM_001025817.4 | NP_001020988.1 | 29 - 30 | 17591 | 71 | ||
| NM_001025818.3 | NP_001020989.1 | 29 - 30 | 17591 | 71 | ||
| NM_001356807.3 | NP_001343717.1 | 30 | 17351 | 71 | ||
| NM_001356808.3 | NP_001343716.1 | 31 | 17792 | 71 | ||
| NM_001356809.3 | NP_001343714.1 | 30 | 19445 | 71 | ||
| NM_001356810.3 | NP_001343718.1 | 30 | 17351 | 71 | ||
| NM_001356811.3 | NP_001343715.1 | 30 | 19445 | 71 | ||
| NM_001356812.3 | NP_001343617.1 | 22 - 23 | 18092 | 71 | ||
| NM_001356813.3 | NP_001343719.1 | 21 - 22 | 15932 | 71 | ||
| NM_001356814.3 | NP_001343711.1 | 22 - 23 | 16373 | 71 | ||
| NM_001356815.3 | NP_001343712.1 | 21 - 22 | 18026 | 71 | ||
| NM_001356816.3 | NP_001343713.1 | 20 - 21 | 16172 | 71 | ||
Set Membership: |
|