Search
Search

erm-1| Gene Symbol: erm-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_059158.7 | NP_491559.1 | 5 - 6 | 714 | 82 | |
| NM_059159.4 | NP_491560.1 | 5 - 6 | 680 | 82 | ||
| GenBank mRNA | AB107268.1 | - | 5 - 6 | 736 | 82 | |
| AB107269.1 | - | 5 - 6 | 702 | 82 | ||
| AY643538.1 | - | 5 - 6 | 704 | 82 | ||
Set Membership: |
|