Search
Search

nhr-31| Gene Symbol: nhr-31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026159.6 | NP_001021330.1 | 8 - 9 | 1651 | 135 | |
| NM_171391.7 | NP_741456.1 | 7 - 8 | 1716 | 135 | ||
| NR_131580.1 | 7 - 8 | 1597 | 135 | |||
| GenBank mRNA | AF273776.1 | - | 7 - 8 | 1706 | 135 | |
Set Membership: |
|