Search
Search

nhr-66| Gene Symbol: nhr-66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028198.5 | NP_001023369.1 | 10 - 11 | 1738 | 91 | |
| NM_001307204.3 | NP_001294133.1 | 7 - 8 | 1318 | 91 | ||
| NM_001307205.2 | NP_001294134.1 | 11 - 12 | 1979 | 91 | ||
| NM_001307206.3 | NP_001294135.1 | 10 - 11 | 1732 | 91 | ||
| NM_001307207.3 | NP_001294136.1 | 8 - 9 | 1498 | 91 | ||
| NM_001380356.1 | NP_001368431.1 | 7 - 8 | 1312 | 91 | ||
| NM_001383138.1 | 11 - 12 | 2066 | 91 | |||
| NM_001383139.1 | 7 - 8 | 1568 | 91 | |||
| NM_171396.7 | NP_741462.1 | 11 - 12 | 1965 | 91 | ||
| NM_171397.8 | NP_741463.1 | 8 - 9 | 1492 | 91 | ||
Set Membership: |
|