Search
Search

unc-62| Gene Symbol: unc-62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029002.4 | NP_001024173.1 | 5 - 6 | 976 | 76 | |
| NM_001313278.3 | NP_001300207.1 | 5 | 865 | 76 | ||
| NM_001380634.1 | NP_001367924.1 | 6 - 7 | 1091 | 76 | ||
| NM_001380635.1 | NP_001367923.1 | 6 - 7 | 1093 | 76 | ||
| NM_001383298.1 | 6 - 7 | 1085 | 76 | |||
| GenBank mRNA | AF427474.1 | - | 6 - 7 | 1113 | 76 | |
| AF427476.1 | - | 6 - 7 | 1296 | 76 | ||
Set Membership: |
|