Search
Search

sma-1| Gene Symbol: sma-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136431.4 | NP_001129903.1 | 2 - 3 | 207 | 70 | |
| NM_001269454.2 | NP_001256383.1 | 2 - 3 | 449 | 70 | ||
| NM_001373124.2 | NP_001360523.1 | 1 | 516 | 70 | ||
| NM_001383415.1 | 2 - 3 | 455 | 70 | |||
| NM_171544.7 | NP_741632.2 | 3 - 4 | 631 | 70 | ||
| GenBank mRNA | AF053496.1 | - | 2 - 3 | 207 | 70 | |
Set Membership: |
|